Assay Details
Gene Symbol: Trmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135876.1 | NP_001129348.1 | 2 - 3 | 314 | 79 | |
XM_006242164.2 | XP_006242226.1 | 2 - 3 | 327 | 79 | ||
XM_008765766.1 | XP_008763988.1 | 1 - 2 | 83 | 79 | ||
GenBank mRNA | BC161991.1 | - | 2 - 3 | 286 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008765766.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
