Assay Details
Gene Symbol: Lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134883.1 | NP_001128355.1 | 5 - 6 | 1659 | 76 | |
XM_006241606.2 | XP_006241668.1 | 4 - 5 | 1555 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
