Assay Details
Gene Symbol: Mkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750385.3 | XP_003750433.1 | 3 - 4 | 5897 | 78 | |
XM_006242130.2 | XP_006242192.1 | 4 - 5 | 954 | 78 | ||
XM_008765806.1 | XP_008764028.1 | 3 - 4 | 1567 | 78 | ||
XM_235497.8 | XP_235497.6 | 4 - 5 | 628 | 78 | ||
GenBank mRNA | AB588919.1 | - | 4 - 5 | 515 | 78 | |
AB588920.1 | - | 3 - 4 | 370 | 78 | ||
AB588921.1 | - | 4 - 5 | 497 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
