Assay Details
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001059906.5 | XP_001059906.3 | 6 - 7 | 1689 | 96 | |
GenBank mRNA | BC169031.1 | - | 6 - 7 | 747 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
