Assay Details
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002016.2 | NP_001002016.2 | 2 - 3 | 712 | 101 | |
GenBank mRNA | DN934590.1 | - | 2 - 3 | 452 | 101 | |
X66870.1 | - | 2 - 3 | 671 | 101 | ||
X99257.1 | - | 2 - 3 | 171 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
