Assay Details
Gene Symbol: Dclk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009691.3 | NP_001009691.3 | 11 - 12 | 2214 | 63 | |
NM_001195832.1 | NP_001182761.1 | 11 - 12 | 2214 | 63 | ||
XM_006232679.2 | XP_006232741.1 | 11 - 12 | 2384 | 63 | ||
XM_006232680.2 | XP_006232742.1 | 11 - 12 | 2381 | 63 | ||
XM_006232681.2 | XP_006232743.1 | 10 - 11 | 2335 | 63 | ||
XM_006232682.2 | XP_006232744.1 | 10 - 11 | 2332 | 63 | ||
XM_006232684.2 | XP_006232746.1 | 11 - 12 | 2384 | 63 | ||
XM_006232685.2 | XP_006232747.1 | 10 - 11 | 2331 | 63 | ||
XM_008761176.1 | XP_008759398.1 | 11 - 12 | 2385 | 63 | ||
XM_008761177.1 | XP_008759399.1 | 11 - 12 | 1279 | 63 | ||
XM_008761178.1 | XP_008759400.1 | 10 - 11 | 2331 | 63 | ||
GenBank mRNA | AY673997.1 | - | 11 - 12 | 1606 | 63 | |
AY673998.1 | - | 11 - 12 | 1686 | 63 | ||
AY674000.1 | - | 11 - 12 | 1606 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
