Assay Details
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020071.2 | NP_064456.2 | 7 - 8 | 1249 | 75 | |
XM_006232524.2 | XP_006232586.1 | 7 - 8 | 1267 | 75 | ||
XM_008761078.1 | XP_008759300.1 | 7 - 8 | 1206 | 75 | ||
GenBank mRNA | AY321323.1 | - | 1245 | 75 | ||
AY325147.1 | - | 1245 | 75 | |||
AY325153.1 | - | 1245 | 75 | |||
BC088102.1 | - | 7 - 8 | 1249 | 75 | ||
FQ209924.1 | - | 7 - 8 | 1256 | 75 | ||
FQ218488.1 | - | 2 - 3 | 333 | 75 | ||
FQ219024.1 | - | 7 - 8 | 1161 | 75 | ||
M27220.1 | - | 4 - 5 | 661 | 75 | ||
U05675.1 | - | 7 - 8 | 1250 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
