Assay Details
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022403.2 | NP_071798.1 | 1 - 2 | 65 | 84 | |
GenBank mRNA | BC089802.1 | - | 1 - 2 | 61 | 7370033 | 84 |
M55167.1 | - | 1 - 2 | 65 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC089802.1, M55167.1, NM_022403.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
