Assay Details
Gene Symbol: Ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106442.1 | NP_001099912.1 | 2 - 3 | 240 | 126 | |
GenBank mRNA | BC168754.1 | - | 2 - 3 | 212 | 126 | |
FQ213377.1 | - | 2 - 3 | 204 | 126 | ||
FQ221799.1 | - | 2 - 3 | 178 | 126 | ||
FQ229540.1 | - | 2 - 3 | 212 | 126 | ||
FQ232929.1 | - | 2 - 3 | 196 | 126 | ||
FQ235190.1 | - | 2 - 3 | 196 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
