Assay Details
Gene Symbol: Cd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012705.1 | NP_036837.1 | 1 - 2 | 111 | 119 | |
XM_006237331.2 | XP_006237393.1 | 3 - 4 | 374 | 119 | ||
XM_006237333.2 | XP_006237395.1 | 2 - 3 | 293 | 119 | ||
XM_008763280.1 | XP_008761502.1 | 1 - 2 | 91 | 119 | ||
GenBank mRNA | M15768.1 | - | 1 - 2 | 111 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
