Assay Details
Gene Symbol: Cd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012705.1 | NP_036837.1 | 2 - 3 | 277 | 159 | |
XM_006237331.2 | XP_006237393.1 | 4 - 5 | 540 | 159 | ||
XM_008763280.1 | XP_008761502.1 | 2 - 3 | 257 | 159 | ||
GenBank mRNA | M15768.1 | - | 2 - 3 | 277 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
