Assay Details
Gene Symbol: Etnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107894.1 | NP_001101364.1 | 1 - 2 | 390 | 156 | |
XM_006237623.1 | XP_006237685.1 | 1 - 2 | 385 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
