Assay Details
Gene Symbol: Etnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107894.1 | NP_001101364.1 | 8 - 9 | 5522 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P019160 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P10213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162229 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P019160 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10213 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P019160 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10213 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P019160 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P10213 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P229633 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10213 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90343 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10859596 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859594 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859593 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859592 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859598 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859597 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859595 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859591 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10859590 | - | - |
Rat Genome 230 2.0 Array | 1371710_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371710_at | - | - |
Rat Genome 230 2.0 Array | 1393151_at | - | - |
Rat Genome 230 2.0 Array | 1393151_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371710_at | - | - |
Affymetrix HT Rat Focus Array | 1393151_at | - | - |
Affymetrix HT Rat Focus Array | 1371710_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393151_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400014590.rn.1 | TC0400002027.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400014589.rn.1 | TC0400002027.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400014592.rn.1 | TC0400002027.rn.1 | RTA TC |
Rat Expression Set 230 | 1371710_at | - | - |
Rat Expression Set 230 | 1371710_PM_at | - | - |
Rat Expression Set 230 | 1393151_at | - | - |
Rat Expression Set 230 | 1393151_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859590 | - | - |
Rat Genome U34 Set | rc_AI058935_at | - | - |
Rat Genome U34 Set | rc_AI232088_at | - | - |
Rat Genome U34 Set | rc_AI176007_at | - | - |
Rat Genome U34 Set | rc_AI145329_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21251678 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530435 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364002 | - | - |
