Assay Details
Gene Symbol: Fgfr1op2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201421.1 | NP_958824.1 | 5 - 6 | 833 | 63 | |
GenBank mRNA | AY426740.1 | - | 5 - 6 | 833 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P156935 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P104154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152727 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156935 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P104154 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P104154 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156935 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P104154 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P156935 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1008329 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P512252 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14054 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47213 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49800 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10859650 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859647 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859654 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859649 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859646 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859651 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859648 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10859645 | - | - |
Rat Genome 230 2.0 Array | 1393829_at | - | - |
Rat Genome 230 2.0 Array | 1393829_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385727_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385727_at | - | - |
Affymetrix HT Rat Focus Array | 1393829_at | - | - |
Affymetrix HT Rat Focus Array | 1385727_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385727_at | - | - |
Affymetrix HT Rat Focus Array | 1393829_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400014777.rn.1 | TC0400002062.rn.1 | RTA TC |
Rat Expression Set 230 | 1393829_at | - | - |
Rat Expression Set 230 | 1393829_PM_at | - | - |
Rat Expression Set 230 | 1385727_PM_at | - | - |
Rat Expression Set 230 | 1385727_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859645 | - | - |
Rat Genome U34 Set | rc_AI639475_at | - | - |
Rat Genome U34 Set | rc_AI179243_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21233244 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373016 | - | - |
RatRef-12 Expression BeadChip | ILMN_69567 | - | - |
