Assay Details
Gene Symbol: Fgfr1op2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237688.2 | XP_006237750.1 | 5 - 6 | 819 | 118 | |
GenBank mRNA | BC087696.1 | - | 5 - 6 | 727 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
