Assay Details
Gene Symbol: Clec7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173386.1 | NP_001166857.1 | 2 - 3 | 239 | 95 | |
GenBank mRNA | AB455106.1 | - | 2 - 3 | 331 | 95 | |
BC166774.1 | - | 2 - 3 | 239 | 95 | ||
GU357485.1 | - | 2 - 3 | 171 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
