Assay Details
Gene Symbol: Clec7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173386.1 | NP_001166857.1 | 4 - 5 | 521 | 89 | |
XM_006237100.2 | XP_006237162.1 | 3 - 4 | 509 | 89 | ||
GenBank mRNA | AB455106.1 | - | 4 - 5 | 613 | 89 | |
AB455107.1 | - | 3 - 4 | 478 | 89 | ||
BC166774.1 | - | 4 - 5 | 521 | 89 | ||
GU357485.1 | - | 4 - 5 | 453 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
