Assay Details
Gene Symbol: LOC100911545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749828.2 | XP_003749876.1 | 3462 | 129 |
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012488.2 | NP_036620.2 | 25 - 26 | 3156 | 129 | |
GenBank mRNA | AY887133.1 | - | 2 - 3 | 220 | 129 | |
BC098922.1 | - | 25 - 26 | 3156 | 129 | ||
J02635.1 | - | 25 - 26 | 3178 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
