Assay Details
Gene Symbol: Far2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_575726.6 | XP_575726.2 | 1 - 2 | 373 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_575726.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
