Assay Details
Gene Symbol: Far2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237697.2 | XP_006237759.1 | 9 - 10 | 2241 | 115 | |
XM_575726.6 | XP_575726.2 | 9 - 10 | 1538 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
