Assay Details
Gene Symbol: Tuba3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040008.1 | NP_001035097.1 | 2 - 3 | 269 | 77 | |
GenBank mRNA | BC079395.1 | - | 2 - 3 | 269 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
