Assay Details
Gene Symbol: Atn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U31777.1 | - | 7 - 8 | 3518 | 117 |
Gene Symbol: LOC100911672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750658.3 | XP_003750706.1 | 7 - 8 | 3508 | 117 | |
XM_006244637.2 | XP_006244699.1 | 7 - 8 | 3494 | 117 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
