Assay Details
Gene Symbol: Etv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037353.1 | NP_001032430.1 | 4 - 5 | 473 | 104 | |
GenBank mRNA | BC105773.1 | - | 4 - 5 | 473 | 7930490 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
