Assay Details
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100496.1 | NP_001093966.1 | 2 - 3 | 230 | 105 | |
XM_006247180.2 | XP_006247242.1 | 3 - 4 | 1034 | 105 | ||
XM_006247181.2 | XP_006247243.1 | 4 - 5 | 341 | 105 | ||
XM_006247182.2 | XP_006247244.1 | 3 - 4 | 290 | 105 | ||
XM_008767968.1 | XP_008766190.1 | 3 - 4 | 386 | 105 | ||
XM_008767969.1 | XP_008766191.1 | 4 - 5 | 308 | 105 | ||
GenBank mRNA | BC166743.1 | - | 2 - 3 | 230 | 7938440 | 105 |
FQ217100.1 | - | 4 - 5 | 538 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
