Assay Details
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100496.1 | NP_001093966.1 | 4 - 5 | 509 | 63 | |
XM_006247180.2 | XP_006247242.1 | 5 - 6 | 1313 | 63 | ||
XM_006247181.2 | XP_006247243.1 | 6 - 7 | 620 | 63 | ||
XM_006247182.2 | XP_006247244.1 | 5 - 6 | 569 | 63 | ||
XM_008767968.1 | XP_008766190.1 | 5 - 6 | 665 | 63 | ||
XM_008767969.1 | XP_008766191.1 | 6 - 7 | 587 | 63 | ||
GenBank mRNA | BC091435.1 | - | 2 - 3 | 235 | 63 | |
BC166743.1 | - | 4 - 5 | 509 | 7938440 | 63 | |
FQ217100.1 | - | 6 - 7 | 817 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
