Assay Details
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246969.2 | XP_006247031.1 | 7 | 2535 | 87 | |
XM_006246970.2 | XP_006247032.1 | 7 | 2532 | 87 | ||
XM_006246973.2 | XP_006247035.1 | 7 | 2772 | 87 | ||
XM_006246974.2 | XP_006247036.1 | 7 | 2362 | 87 | ||
XM_006246975.2 | XP_006247037.2 | 8 | 3343 | 87 | ||
XM_006246977.2 | XP_006247039.1 | 7 | 3251 | 87 | ||
XM_006246978.2 | XP_006247040.1 | 7 | 2769 | 87 | ||
XM_006246979.2 | XP_006247041.1 | 7 | 2358 | 87 | ||
XM_008767954.1 | XP_008766176.1 | 8 | 3340 | 87 | ||
XM_008767955.1 | XP_008766177.1 | 8 | 3525 | 87 | ||
XM_008767956.1 | XP_008766178.1 | 7 | 3260 | 87 | ||
GenBank mRNA | FQ210900.1 | - | 7 | 2243 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
