Assay Details
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246974.2 | XP_006247036.1 | 1 - 2 | 194 | 111 | |
XM_006246979.2 | XP_006247041.1 | 1 - 2 | 193 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006246974.2, XM_006246979.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
