Assay Details
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246969.2 | XP_006247031.1 | 3 - 4 | 966 | 64 | |
XM_006246970.2 | XP_006247032.1 | 3 - 4 | 963 | 64 | ||
XM_006246973.2 | XP_006247035.1 | 3 - 4 | 1203 | 64 | ||
XM_006246974.2 | XP_006247036.1 | 3 - 4 | 793 | 64 | ||
XM_006246975.2 | XP_006247037.2 | 4 - 5 | 1774 | 64 | ||
XM_006246977.2 | XP_006247039.1 | 3 - 4 | 1682 | 64 | ||
XM_006246978.2 | XP_006247040.1 | 3 - 4 | 1200 | 64 | ||
XM_006246979.2 | XP_006247041.1 | 3 - 4 | 789 | 64 | ||
XM_008767954.1 | XP_008766176.1 | 4 - 5 | 1771 | 64 | ||
XM_008767955.1 | XP_008766177.1 | 4 - 5 | 1956 | 64 | ||
GenBank mRNA | AY157969.1 | - | 2 - 3 | 585 | 64 | |
FQ210900.1 | - | 3 - 4 | 674 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
