Assay Details
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246969.2 | XP_006247031.1 | 4 - 5 | 1049 | 73 | |
XM_006246970.2 | XP_006247032.1 | 4 - 5 | 1046 | 73 | ||
XM_006246973.2 | XP_006247035.1 | 4 - 5 | 1286 | 73 | ||
XM_006246974.2 | XP_006247036.1 | 4 - 5 | 876 | 73 | ||
XM_006246975.2 | XP_006247037.2 | 5 - 6 | 1857 | 73 | ||
XM_006246977.2 | XP_006247039.1 | 4 - 5 | 1765 | 73 | ||
XM_006246978.2 | XP_006247040.1 | 4 - 5 | 1283 | 73 | ||
XM_006246979.2 | XP_006247041.1 | 4 - 5 | 872 | 73 | ||
XM_008767954.1 | XP_008766176.1 | 5 - 6 | 1854 | 73 | ||
XM_008767955.1 | XP_008766177.1 | 5 - 6 | 2039 | 73 | ||
GenBank mRNA | AY157969.1 | - | 3 - 4 | 668 | 73 | |
FQ210900.1 | - | 4 - 5 | 757 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
