Assay Details
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004068.1 | NP_001004068.1 | 3 - 4 | 893 | 69 | |
XM_008767956.1 | XP_008766178.1 | 4 - 5 | 1776 | 69 | ||
GenBank mRNA | BC079216.1 | - | 3 - 4 | 893 | 7131890 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
