Assay Details
Gene Symbol: Il9r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017021.1 | NP_058717.1 | 2 - 3 | 149 | 99 | |
XM_006245901.2 | XP_006245963.1 | 1 - 2 | 491 | 99 | ||
XM_006245903.2 | XP_006245965.1 | 2 - 3 | 511 | 99 | ||
GenBank mRNA | L36459.1 | - | 2 - 3 | 149 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
