Assay Details
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030034.1 | NP_001025205.1 | 1 - 2 | 109 | 50 | |
GenBank mRNA | BC099777.1 | - | 1 - 2 | 109 | 7933232 | 50 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC099777.1, NM_001030034.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
