Assay Details
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030034.1 | NP_001025205.1 | 4 - 5 | 590 | 138 | |
XM_006245977.2 | XP_006246039.1 | 4 - 5 | 4408 | 138 | ||
XM_008767578.1 | XP_008765800.1 | 5 - 6 | 3525 | 138 | ||
XM_008767579.1 | XP_008765801.1 | 5 - 6 | 3400 | 138 | ||
XM_008767580.1 | XP_008765802.1 | 4 - 5 | 1510 | 138 | ||
GenBank mRNA | BC099777.1 | - | 4 - 5 | 590 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
