Assay Details
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030034.1 | NP_001025205.1 | 6 - 7 | 928 | 71 | |
XM_006245977.2 | XP_006246039.1 | 6 - 7 | 4746 | 71 | ||
XM_008767578.1 | XP_008765800.1 | 7 - 8 | 3863 | 71 | ||
XM_008767579.1 | XP_008765801.1 | 7 - 8 | 3738 | 71 | ||
XM_008767580.1 | XP_008765802.1 | 6 - 7 | 1848 | 71 | ||
GenBank mRNA | BC099777.1 | - | 6 - 7 | 928 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
