Assay Details
Gene Symbol: Gabra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021841.1 | NP_068613.1 | 6 - 7 | 669 | 119 | |
XM_006246128.2 | XP_006246190.1 | 6 - 7 | 818 | 119 | ||
XM_008767636.1 | XP_008765858.1 | 4 - 5 | 740 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
