Assay Details
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031982.1 | NP_114188.1 | 10 - 11 | 1631 | 68 | |
XM_008767882.1 | XP_008766104.1 | 9 - 10 | 1638 | 68 | ||
GenBank mRNA | AB015231.3 | - | 882 | 68 | ||
AB040873.1 | - | 9 - 10 | 1551 | 68 | ||
AB041029.1 | - | 8 - 9 | 1371 | 68 | ||
AF029310.1 | - | 10 - 11 | 1631 | 68 | ||
AF158248.2 | - | 4 - 5 | 964 | 68 | ||
LC008303.1 | - | 5 - 6 | 871 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
