Assay Details
Gene Symbol: Coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006954.1 | NP_001006955.1 | 5 - 6 | 1325 | 127 | |
XM_006247276.2 | XP_006247338.1 | 5 - 6 | 1706 | 127 | ||
XM_008767975.1 | XP_008766197.1 | 6 - 7 | 1527 | 127 | ||
GenBank mRNA | BC083781.1 | - | 5 - 6 | 1325 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
