Assay Details
Gene Symbol: Ap2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080583.1 | NP_542150.1 | 14 - 15 | 2132 | 88 | |
XM_008767935.1 | XP_008766157.1 | 15 - 16 | 2889 | 88 | ||
XM_008767936.1 | XP_008766158.1 | 14 - 15 | 2103 | 88 | ||
XM_008767937.1 | XP_008766159.1 | 13 - 14 | 2018 | 88 | ||
XM_008767938.1 | XP_008766160.1 | 13 - 14 | 2035 | 88 | ||
GenBank mRNA | BC103481.1 | - | 15 - 16 | 2101 | 88 | |
M77246.1 | - | 14 - 15 | 2132 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
