Assay Details
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011890.1 | NP_001011890.1 | 2 - 3 | 241 | 77 | |
XM_006245931.2 | XP_006245993.1 | 2 - 3 | 685 | 77 | ||
XM_006245932.2 | XP_006245994.1 | 3 - 4 | 384 | 77 | ||
GenBank mRNA | BC079014.1 | - | 2 - 3 | 241 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
