Assay Details
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011890.1 | NP_001011890.1 | 5 - 6 | 689 | 113 | |
XM_006245931.2 | XP_006245993.1 | 5 - 6 | 1133 | 113 | ||
XM_006245932.2 | XP_006245994.1 | 6 - 7 | 832 | 113 | ||
XM_006245933.2 | XP_006245995.1 | 4 - 5 | 628 | 113 | ||
GenBank mRNA | BC079014.1 | - | 5 - 6 | 689 | 7113485 | 113 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
