Assay Details
Gene Symbol: Foxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100648.1 | NP_001094118.1 | 5 - 6 | 938 | 68 | |
GenBank mRNA | S80120.1 | - | 5 - 6 | 938 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
