Assay Details
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012528.1 | NP_036660.1 | 4 - 5 | 423 | 80 | |
XM_008767770.1 | XP_008765992.1 | 4 - 5 | 470 | 80 | ||
GenBank mRNA | X74833.1 | - | 4 - 5 | 423 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
