Assay Details
Gene Symbol: Aurkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053749.1 | NP_446201.1 | 6 - 7 | 730 | 91 | |
XM_006246569.2 | XP_006246631.1 | 6 - 7 | 748 | 91 | ||
XM_006246570.2 | XP_006246632.1 | 5 - 6 | 635 | 91 | ||
XM_008767757.1 | XP_008765979.1 | 6 - 7 | 645 | 91 | ||
XM_008767758.1 | XP_008765980.1 | 6 - 7 | 591 | 91 | ||
GenBank mRNA | BC097297.1 | - | 6 - 7 | 648 | 7454285 | 91 |
D89731.1 | - | 6 - 7 | 730 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
