Assay Details
Gene Symbol: Acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034951.1 | NP_001030123.1 | 14 - 15 | 1853 | 129 | |
GenBank mRNA | BC099826.1 | - | 14 - 15 | 1853 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
