Assay Details
Gene Symbol: Acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034951.1 | NP_001030123.1 | 15 - 16 | 1927 | 133 | |
GenBank mRNA | BC099826.1 | - | 15 - 16 | 1927 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P458494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7194 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P458494 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P458494 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P458494 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90467 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57301 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10314 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10746400 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746414 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746402 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746405 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746403 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746415 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746413 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746406 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746408 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746409 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746412 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746411 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746404 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746407 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746410 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746401 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10746399 | - | - |
Rat Genome 230 2.0 Array | 1392952_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392952_at | - | - |
Affymetrix HT Rat Focus Array | 1392952_at | - | - |
Affymetrix HT Rat Focus Array | 1392952_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000032097.rn.1 | TC1000003595.rn.1 | RTA TC |
Rat Expression Set 230 | 1392952_PM_at | - | - |
Rat Expression Set 230 | 1392952_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746399 | - | - |
Rat Genome U34 Set | rc_AA955369_s_at | - | - |
Rat Genome U34 Set | rc_AI010027_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22083284 | - | - |
ABI Rat Genome Survey Microarray | 22344023 | - | - |
