Assay Details
Gene Symbol: Lgals9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012977.1 | NP_037109.1 | 1 - 2 | 125 | 102 | |
GenBank mRNA | BC061566.1 | - | 1 - 2 | 86 | 102 | |
U67958.1 | - | 1 - 2 | 125 | 102 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC061566.1, NM_012977.1, U67958.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
