Assay Details
Gene Symbol: Cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024488.1 | NP_077814.1 | 11 - 12 | 1160 | 75 | |
GenBank mRNA | AF177476.1 | - | 11 - 12 | 1160 | 75 | |
BC081793.1 | - | 10 - 11 | 1082 | 7190258 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
