Assay Details
Gene Symbol: Pemt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013003.1 | NP_037135.1 | 1 - 2 | 78 | 59 | |
GenBank mRNA | BC091162.1 | - | 1 - 2 | 118 | 59 | |
L14441.1 | - | 1 - 2 | 78 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091162.1, L14441.1, NM_013003.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
