Assay Details
Gene Symbol: Pemt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013003.1 | NP_037135.1 | 3 - 4 | 298 | 68 | |
GenBank mRNA | BC091162.1 | - | 3 - 4 | 338 | 7325819 | 68 |
L14441.1 | - | 3 - 4 | 298 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
