Assay Details
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106992.1 | NP_001100462.1 | 7 - 8 | 1758 | 86 | |
XM_006245978.2 | XP_006246040.1 | 8 - 9 | 3156 | 86 | ||
XM_006245979.2 | XP_006246041.1 | 7 - 8 | 3105 | 86 | ||
XM_006245980.2 | XP_006246042.1 | 6 - 7 | 3081 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P038172 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P033698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60211 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P033698 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038172 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038172 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P033698 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P033698 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P038172 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P361857 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P756696 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16651 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P732939 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53969 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10741819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741815 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10741814 | - | - |
Rat Genome 230 2.0 Array | 1372223_at | - | - |
Rat Genome 230 2.0 Array | 1393294_at | - | - |
Rat Genome 230 2.0 Array | 1372223_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393294_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000022478.rn.1 | TC1000002473.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000022480.rn.1 | TC1000002473.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000022479.rn.1 | TC1000002473.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000022477.rn.1 | TC1000002473.rn.1 | RTA TC |
Rat Expression Set 230 | 1372223_at | - | - |
Rat Expression Set 230 | 1372223_PM_at | - | - |
Rat Expression Set 230 | 1393294_PM_at | - | - |
Rat Expression Set 230 | 1393294_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741814 | - | - |
Rat Genome U34 Set | rc_AA818407_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20837062 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371786 | - | - |
